3-(Trifluoromethoxy)thiophenol is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(Trifluoromethoxy)thiophenol is used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
V.G.Nenajdenko; A.V.Sanin; A.V.Churakov; J.A.K.Howard; E.S.Balenkova. Reaction of trifluoromethyl-containing enones with thiophenol derivatives. Chemistry of Heterocyclic Compounds. 1999, 35, (5),549-556
Jenny B.Waern; Peter Turner; Margaret M.Harding. Synthesis and hydrolysis of thiol derivatives of molybdocene dichloride incorporating electron-withdrawing substituents. Organometallics. 2006, 25, (14),3417-3421