Thermo Scientific Chemicals

7-Bromo-1-chloroisoquinoline, 97%, Thermo Scientific Chemicals

Catalog number: L19946.06
5 g, Each
Thermo Scientific Chemicals

7-Bromo-1-chloroisoquinoline, 97%, Thermo Scientific Chemicals

Catalog number: L19946.06
5 g, Each
Quantity
Catalog number: L19946.06
also known as L19946-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
215453-51-3
IUPAC Name
7-bromo-1-chloroisoquinoline
Molecular Formula
C9H5BrClN
InChI Key
UMSWWSIVPWVJOX-UHFFFAOYSA-N
SMILES
ClC1=NC=CC2=CC=C(Br)C=C12
Appearance (Color)
Cream or yellow
Water Content (Karl Fischer Titration)
< 1.5%
Assay (GC)
> 96.0%
Form
crystalline powder
Assay (HPLC)
> 96.0%

Description

7-Bromo-1-chloroisoquinoline as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-1-chloroisoquinoline as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text