Thermo Scientific Chemicals

N-Boc-ethylenediamine, 98%, may cont up to 5% tert-butanol, Thermo Scientific Chemicals

Catalog number: L19947.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-ethylenediamine, 98%, may cont up to 5% tert-butanol, Thermo Scientific Chemicals

Catalog number: L19947.06
5 g, Each
Quantity
Catalog number: L19947.06
also known as L19947-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57260-73-8
IUPAC Name
tert-butyl N-(2-aminoethyl)carbamate
Molecular Formula
C7H16N2O2
InChI Key
AOCSUUGBCMTKJH-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NCCN
Appearance (Color)
Clear, colorless to pale yellow
Form
Liquid
Assay (Non-aqueous acid-base Titration)
>97.5 to <102.5% (excluding tert-Butanol)
Assay (GC)
>97.5% (excluding tert-Butanol)
Impurity content
(tert-Butanol) : ≤ 5.0%

Description

N-Boc-ethylenediamine is used in the synthesis of thyronamine derivatives. It is also used in the preparation of (2-isothiocyanato-ethyl)-carbamic acid tert-butyl ester by reacting with carbon disulfide. Further, it is used in both oligonucleotide and peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-ethylenediamine is used in the synthesis of thyronamine derivatives. It is also used in the preparation of (2-isothiocyanato-ethyl)-carbamic acid tert-butyl ester by reacting with carbon disulfide. Further, it is used in both oligonucleotide and peptide synthesis.

Solubility
Miscible with methanol and chloroform. Slightly miscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text