Thermo Scientific Chemicals

2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L19952.MD
250 mg, Each
Thermo Scientific Chemicals

2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L19952.MD
250 mg, Each
Quantity
Catalog number: L19952.MD
also known as L19952-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
681812-07-7
IUPAC Name
[2,6-bis(trifluoromethyl)phenyl]boronic acid
Molecular Formula
C8H5BF6O2
InChI Key
WAMPGNNEOZGBHR-UHFFFAOYSA-N
SMILES
OB(O)C1=C(C=CC=C1C(F)(F)F)C(F)(F)F
Appearance (Color)
Pale cream to cream to yellow
Form
Powder
Assay (HPLC)
≥96.0%
Proton NMR
Conforms to structure

Description

2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text