Thermo Scientific Chemicals

1-Octylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L19964.06
5 g, Each
Thermo Scientific Chemicals

1-Octylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: L19964.06
5 g, Each
Quantity
Catalog number: L19964.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28741-08-4
IUPAC Name
octylboronic acid
Molecular Formula
C8H19BO2
InChI Key
GKFRVXOKPXCXAK-UHFFFAOYSA-N
SMILES
CCCCCCCCB(O)O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0%
Proton NMR
Conforms to structure

Description

1-Octylboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text