1-Octylboronic acid, 97%, Thermo Scientific Chemicals
1-Octylboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Octylboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number L19964.14
Price (USD)/ Each
575.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
575.00
Add to cart
1-Octylboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberL19964.14
Price (USD)/ Each
575.00
-
Add to cart
Chemical Identifiers
CAS28741-08-4
IUPAC Nameoctylboronic acid
Molecular FormulaC8H19BO2
InChI KeyGKFRVXOKPXCXAK-UHFFFAOYSA-N
SMILESCCCCCCCCB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0%
Proton NMRConforms to structure
1-Octylboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
Kenneth D.Clevenger; Rui Wu; Dali Liu; Walter Fast. n-Alkylboronic acid inhibitors reveal determinants of ligand specificity in the quorum-quenching and siderophore biosynthetic enzyme PvdQ. Biochemistry. 2014, 53,(42), 6679-6686.