4-Iodobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals
4-Iodobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Iodobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L19967.06
also known as L19967-06
Price (USD)/ Each
261.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
261.00
Add to cart
4-Iodobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberL19967.06
Price (USD)/ Each
261.00
-
Add to cart
Chemical Identifiers
CAS59528-27-7
IUPAC Name1-(4-iodophenyl)methanamine hydrochloride
Molecular FormulaC7H9ClIN
InChI KeyGBJMURRFWZREHE-UHFFFAOYSA-N
SMILESCl.NCC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
4-Iodobenzylamine hydrochloride is used to prepare cyclic amide by reacting with methyl 4-bromomethyl-3-methoxycarbonylcinnamate in the presence of triethylamine. It is also used in pharmaceuticals, agrochemicals and dye industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzylamine hydrochloride is used to prepare cyclic amide by reacting with methyl 4-bromomethyl-3-methoxycarbonylcinnamate in the presence of triethylamine. It is also used in pharmaceuticals, agrochemicals and dye industries.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Strack, M.; Langklotz, S.; Bandow, J. E.; Metzler-Nolte, N.; Albada, H. B. Silyl-Based Alkyne-Modifying Linker for the Preparation of C-Terminal Acetylene-Derivatized Protected Peptides. J. Org. Chem. 2012, 77 (22), 9954-9958.
  2. Wijtmans, M.; Verzijl, D.; Bergmans, S.; Lai, M.; Bosch, L.; Smit, M. J.; de Esch, I. J. P.; Leurs, R. CXCR3 antagonists: Quaternary ammonium salts equipped with biphenyl- and polycycloaliphatic-anchors. Bioorg. Med. Chem. 2011, 19 (11), 3384-3393.