1-(4-Nitrophenyl)glycerol, 99%, Thermo Scientific Chemicals
1-(4-Nitrophenyl)glycerol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Nitrophenyl)glycerol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L19968.06
also known as L19968-06
Price (USD)/ Each
627.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
627.00
Add to cart
1-(4-Nitrophenyl)glycerol, 99%, Thermo Scientific Chemicals
Catalog numberL19968.06
Price (USD)/ Each
627.00
-
Add to cart
Chemical Identifiers
CAS2207-68-3
IUPAC Name1-(4-nitrophenyl)propane-1,2,3-triol
Molecular FormulaC9H11NO5
InChI KeyIUZVZBIQZKBWCC-UHFFFAOYNA-N
SMILESOCC(O)C(O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale yellow
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤0.5%
Melting Point (clear melt)94.0-100.0?C
Assay (HPLC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only