Thermo Scientific Chemicals

5-Chloro-2-furaldehyde, 98%, stab. with 2% ethanol, Thermo Scientific Chemicals

Catalog number: L19971.03
1 g, Each
Thermo Scientific Chemicals

5-Chloro-2-furaldehyde, 98%, stab. with 2% ethanol, Thermo Scientific Chemicals

Catalog number: L19971.03
1 g, Each
Quantity
Catalog number: L19971.03
also known as L19971-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21508-19-0
IUPAC Name
5-chlorofuran-2-carbaldehyde
Molecular Formula
C5H3ClO2
InChI Key
DGAUAVDWXYXXGQ-UHFFFAOYSA-N
SMILES
ClC1=CC=C(O1)C=O
Appearance (Color)
White to cream or pale yellow
Form
crystalline powder
Assay (GC)
> 97.5%
Free acid (titration)
< 1.0%

Description

(+)-1-Deoxynojirimycin hydrochloride interferes with normal processing of N-linked glycoproteins, producing reduced amounts of complex type oligosaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-1-Deoxynojirimycin hydrochloride interferes with normal processing of N-linked glycoproteins, producing reduced amounts of complex type oligosaccharides.

Solubility
Insoluble in water.

Notes
Air
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text