Thermo Scientific Chemicals

2,6-Dichloronicotinic acid, 97%, Thermo Scientific Chemicals

Catalog number: L20002.03
1 g, Each
Thermo Scientific Chemicals

2,6-Dichloronicotinic acid, 97%, Thermo Scientific Chemicals

Catalog number: L20002.03
1 g, Each
Quantity
Catalog number: L20002.03
also known as L20002-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38496-18-3
IUPAC Name
2,6-dichloropyridine-3-carboxylic acid
Molecular Formula
C6H3Cl2NO2
InChI Key
AJPKQSSFYHPYMH-UHFFFAOYSA-N
SMILES
OC(=O)C1=C(Cl)N=C(Cl)C=C1
Appearance (Color)
White to pale cream to cream to yellow to brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text