Thermo Scientific Chemicals

4-Amino-2-bromopyridine, 97%, Thermo Scientific Chemicals

Catalog number: L20007.06
5 g, Each
Thermo Scientific Chemicals

4-Amino-2-bromopyridine, 97%, Thermo Scientific Chemicals

Catalog number: L20007.06
5 g, Each
Quantity
Catalog number: L20007.06
also known as L20007-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7598-35-8
Appearance (Color)
Pale cream to cream
Form
Powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
93.0-102.0?C

Description

4-Amino-2-bromopyridine is used as medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2-bromopyridine is used as medical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text