Thermo Scientific Chemicals

2-Bromo-6-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: L20018.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-6-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: L20018.06
5 g, Each
Quantity
Catalog number: L20018.06
also known as L20018-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5315-25-3
IUPAC Name
2-bromo-6-methylpyridine
Molecular Formula
C6H6BrN
InChI Key
SOHDPICLICFSOP-UHFFFAOYSA-N
SMILES
CC1=CC=CC(Br)=N1
Identification (FTIR)
Conforms
Refractive Index
1.5605-1.5655 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%

Description

6-Bromo-2-picoline is a halogenated pyridine derivative used is a building block in the preparation of nitrogen containing heterocyclic compounds. It may used in following synthesis 6,6?-dimethyl-2,2?-bipyridine, 6-methyl-2-pyridyl-2-pyridylmethanone, 2-methyl-6-(trimethylsilyl)-pyridine and N,N’-bis-(6-methylpyrid-2-yl)-(1R,2R)-1,2- diaminocyclohexane (cydiampy).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-2-picoline is a halogenated pyridine derivative used is a building block in the preparation of nitrogen containing heterocyclic compounds. It may used in following synthesis 6,6′-dimethyl-2,2′-bipyridine, 6-methyl-2-pyridyl-2-pyridylmethanone, 2-methyl-6-(trimethylsilyl)-pyridine and N,N’-bis-(6-methylpyrid-2-yl)-(1R,2R)-1,2- diaminocyclohexane (cydiampy).

Solubility
Soluble in chloroform, ethyl aceate. Not miscible or difficult to mix with water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text