4-Chloronicotinic acid, 96%, Thermo Scientific Chemicals
4-Chloronicotinic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloronicotinic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20029.03
also known as L20029-03
Price (USD)/ Each
83.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
83.00
Add to cart
4-Chloronicotinic acid, 96%, Thermo Scientific Chemicals
Catalog numberL20029.03
Price (USD)/ Each
83.00
-
Add to cart
Chemical Identifiers
CAS10177-29-4
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)Pale cream to cream to yellow to pale brown
Assay (HPLC)≥96.0%
4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.

Solubility
Soluble in organic solvents at 20 deg C

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers
RUO – Research Use Only

General References:

  1. Edward C. Taylor.; John S. Driscoll. Pyridine-1-oxides. V. 4-Substituted Nicotinic Acid-1-oxides1. J. Am. Chem. Soc. 1960, 82, (12), 3141-3143.
  2. Edward C. Taylor.; Wendell A. Ehrhart. A Convenient Synthesis of Formamidine and Acetamidine Acetate. J. Am. Chem. Soc. 1960, 82, (12), 3138-3141.