Thermo Scientific Chemicals

4-Chloronicotinic acid, 96%, Thermo Scientific Chemicals

Catalog number: L20029.06
5 g, Each
Thermo Scientific Chemicals

4-Chloronicotinic acid, 96%, Thermo Scientific Chemicals

Catalog number: L20029.06
5 g, Each
Quantity
Catalog number: L20029.06
also known as L20029-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10177-29-4
Form
Powder or lumps
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
Pale cream to cream to yellow to pale brown
Assay (HPLC)
≥96.0%

Description

4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.

Solubility
Soluble in organic solvents at 20 deg C

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text