3-Fluoropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
3-Fluoropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20042.03
also known as L20042-03
Price (USD)/ Each
50.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
50.00
Add to cart
3-Fluoropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog numberL20042.03
Price (USD)/ Each
50.00
-
Add to cart
Chemical Identifiers
CAS393-53-3
IUPAC Name3-fluoropyridine-4-carboxylic acid
Molecular FormulaC6H4FNO2
InChI KeyPOLXLLIQWDBJMD-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=NC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only