2,3,6-Trifluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3,6-Trifluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number L20053.03
also known as L20053-03
Price (USD)/ Each
299.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
299.00
Add to cart
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Catalog numberL20053.03
Price (USD)/ Each
299.00
-
Add to cart
Chemical Identifiers
CAS84940-46-5
IUPAC Name2,3,6-trifluoro-4-(trifluoromethyl)pyridine
Molecular FormulaC6HF6N
InChI KeyDODFHNWFNCGXOW-UHFFFAOYSA-N
SMILESFC1=NC(F)=C(F)C(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.3745-1.3795 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only