2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
RUO – Research Use Only
Gayatri Sarmah.; Utpal Bora. Simple aminobenzoic acid promoted palladium catalyzed room temperature Suzuki-Miyaura cross-coupling reaction in aqueous media. Tetrahedron Letters. 2015, 56, (22), 2906-2909.