4-Chloro-3-(trifluoromethyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
4-Chloro-3-(trifluoromethyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-3-(trifluoromethyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L20105.06
also known as L20105-06
Price (USD)/ Each
319.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
319.00
Add to cart
4-Chloro-3-(trifluoromethyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
Catalog numberL20105.06
Price (USD)/ Each
319.00
-
Add to cart
Chemical Identifiers
CAS176976-42-4
IUPAC Name[4-chloro-3-(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC7H5BClF3O2
InChI KeyXHKNQBNGLMOTDB-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Cl)C(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
Assay (Aqueous acid-base Titration)≥95.0%
Appearance (Color)White to cream
Assay (HPLC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only