2-Fluoropyridine-3-boronic acid, 97%, Thermo Scientific Chemicals
2-Fluoropyridine-3-boronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoropyridine-3-boronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L20108.06
also known as L20108-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS174669-73-9
IUPAC Name(2-fluoropyridin-3-yl)boronic acid
Molecular FormulaC5H5BFNO2
InChI KeyYUHZIUAREWNXJT-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CN=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥96.0%
Proton NMRConforms to structure
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only