3-Aminophenylacetylene, 98%, Thermo Scientific Chemicals
3-Aminophenylacetylene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminophenylacetylene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L20124.06
also known as L20124-06
Price (USD)/ Each
83.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
83.40
Add to cart
3-Aminophenylacetylene, 98%, Thermo Scientific Chemicals
Catalog numberL20124.06
Price (USD)/ Each
83.40
-
Add to cart
Chemical Identifiers
CAS54060-30-9
IUPAC Name3-ethynylaniline
Molecular FormulaC8H7N
InChI KeyNNKQLUVBPJEUOR-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow to orange to brown
Refractive Index1.6145-1.6185 @ 20°C
Assay (GC)≥97.5%
FormLiquid
Identification (FTIR)Conforms
It is used as Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as Pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heavy metal powders.
RUO – Research Use Only
Anatoli Onopchenko; Edward T. Sabourin; Charles M. Selwitz. Selective catalytic hydrogenation of aromatic nitro groups in the presence of acetylenes. Synthesis of (3-aminophenyl)acetylene via hydrogenation of (3-nitrophenyl)acetylene over cobalt polysulfide and ruthenium sulfide catalysts. J. Org. Chem. 1979, 44(21), 3671-3674