4-Aminopiperidine, 95%, Thermo Scientific Chemicals
4-Aminopiperidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Aminopiperidine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L20127.06
also known as L20127-06
Price (USD)/ Each
59.65
Online exclusive
65.80 
Save 6.15 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
59.65
Online exclusive
65.80 
Save 6.15 (9%)
Add to cart
4-Aminopiperidine, 95%, Thermo Scientific Chemicals
Catalog numberL20127.06
Price (USD)/ Each
59.65
Online exclusive
65.80 
Save 6.15 (9%)
-
Add to cart
Chemical Identifiers
CAS13035-19-3
IUPAC Namepiperidin-4-amine
Molecular FormulaC5H12N2
InChI KeyBCIIMDOZSUCSEN-UHFFFAOYSA-N
SMILESNC1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or low melting solid
Assay (GC)≥94.0%
Appearance (Color)Clear colorless to yellow to orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only