Thermo Scientific Chemicals

3H-1,2-Benzodithiol-one 1,1-dioxide, 98%, Thermo Scientific Chemicals

Catalog number: L20129.MD
250 mg, Each
Thermo Scientific Chemicals

3H-1,2-Benzodithiol-one 1,1-dioxide, 98%, Thermo Scientific Chemicals

Catalog number: L20129.MD
250 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L20129.MD
also known as L20129-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66304-01-6
IUPAC Name
3H-1λ⁶,2-benzodithiole-1,1,3-trione
Molecular Formula
C7H4O3S2
InChI Key
JUDOLRSMWHVKGX-UHFFFAOYSA-N
SMILES
O=C1SS(=O)(=O)C2=CC=CC=C12
Identification (FTIR)
Conforms
Appearance (Color)
Colorless to white
Water Content (Karl Fischer Titration)
≤0.5%
Melting Point (clear melt)
100-107?C
Assay (HPLC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text