Thermo Scientific Chemicals

(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%, Thermo Scientific Chemicals

Catalog number: L20131.03
1 g, Each
Thermo Scientific Chemicals

(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%, Thermo Scientific Chemicals

Catalog number: L20131.03
1 g, Each
Quantity
Catalog number: L20131.03
also known as L20131-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
113451-59-5
IUPAC Name
tert-butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
Molecular Formula
C10H18N2O2
InChI Key
UXAWXZDXVOYLII-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)N1CC2CC1CN2
Form
powder
Appearance (Color)
White to pale cream
Assay (Silylated GC)
>96.0% (sum of enantiomers)
Optical Rotation
-46? ? 2? (c=1 chloroform)

Description

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air, carbon dioxide, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text