(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%, Thermo Scientific Chemicals
(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L20131.06
also known as L20131-06
Price (USD)/ Each
604.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
604.00
Add to cart
(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%, Thermo Scientific Chemicals
Catalog numberL20131.06
Price (USD)/ Each
604.00
-
Add to cart
Chemical Identifiers
CAS113451-59-5
IUPAC Nametert-butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
Molecular FormulaC10H18N2O2
InChI KeyUXAWXZDXVOYLII-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CC2CC1CN2
View more
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Appearance (Color)White to pale cream
Assay (Silylated GC)>96.0% (sum of enantiomers)
Optical Rotation-46? ? 2? (c=1 chloroform)
It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air, carbon dioxide, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
RUO – Research Use Only