Thermo Scientific Chemicals

4-Bromo-2-fluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L20142.06
5 g, Each
Thermo Scientific Chemicals

4-Bromo-2-fluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: L20142.06
5 g, Each
Quantity
Catalog number: L20142.06
also known as L20142-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
151982-51-3
IUPAC Name
4-bromo-2-fluorobenzoyl chloride
Molecular Formula
C7H3BrClFO
InChI Key
PCFIABOQFAFDAU-UHFFFAOYSA-N
SMILES
FC1=CC(Br)=CC=C1C(Cl)=O
Appearance (Color)
Colorless to white to pale yellow
Assay (GC)
≥97.5%
Form
Fused solid or clear liquid as melt
Refractive Index
1.5860-1.5900 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text