2-Chloro-5-methoxyaniline, 98+%, Thermo Scientific Chemicals
2-Chloro-5-methoxyaniline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-5-methoxyaniline, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L20162.06
also known as L20162-06
Price (USD)/ Each
63.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
63.90
Add to cart
2-Chloro-5-methoxyaniline, 98+%, Thermo Scientific Chemicals
Catalog numberL20162.06
Price (USD)/ Each
63.90
-
Add to cart
Chemical Identifiers
CAS2401-24-3
IUPAC Name2-chloro-5-methoxyaniline
Molecular FormulaC7H8ClNO
InChI KeyGBOUQGUQUUPGLO-UHFFFAOYSA-N
SMILESCOC1=CC=C(Cl)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5845-1.5885 @ 20?C
Appearance (Color)Clear pale yellow to pale orange to brown
Assay (GC)≥98.0%
2-Chloro-5-methoxyaniline is used in the preparation of 4-amino-5-chloro-2-methoxyphenyl thiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-methoxyaniline is used in the preparation of 4-amino-5-chloro-2-methoxyphenyl thiocyanate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Chen, S.; Lu, G.; Cai, C. Iridium-catalyzed transfer hydrogenation of nitroarenes to anilines. New J. Chem. 2015, 39 (7), 5360-5365.
  2. Morisson-Iveson, V.; Wadsworth, H.; Passmore, J.; Ewan, A.; Nilsen, S.; Thaning, M.; Trigg, W. An improved, regioselective synthesis of the radiolabelling precursor for the translocator protein targeting positron emission tomography imaging radiotracer [18 F] GE-180. Tetrahedron Lett. 2014, 55 (37), 5141-5143.