Thermo Scientific Chemicals

(1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide, 98%, Thermo Scientific Chemicals

Catalog number: L20170.03
1 g, Each
Thermo Scientific Chemicals

(1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide, 98%, Thermo Scientific Chemicals

Catalog number: L20170.03
1 g, Each
Quantity
Catalog number: L20170.03
also known as L20170-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
132747-20-7
IUPAC Name
2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Molecular Formula
C5H12Br2N2
InChI Key
ISYQWKOXKGJREA-UHFFFAOYNA-N
SMILES
Br.Br.C1NC2CNC1C2
Appearance (Color)
White to pale brown or purple
Assay (Titration ex Bromide)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Optical Rotation
22.4 ± 0.4? (C=1 in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text