Thermo Scientific Chemicals

4-(Dimethylamino)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: L20176.14
25 g, Each
Thermo Scientific Chemicals

4-(Dimethylamino)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: L20176.14
25 g, Each
Quantity
Catalog number: L20176.14
also known as L20176-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50533-97-6
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4735-1.4785 @ 20?C

Description

4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.

Notes
Incompatible with air, water, oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text