Thermo Scientific Chemicals

2,2-Dimethyl-1,3-dioxan-5-one, tech. 90%, Thermo Scientific Chemicals

Catalog number: L20177.03
1 g, Each
Thermo Scientific Chemicals

2,2-Dimethyl-1,3-dioxan-5-one, tech. 90%, Thermo Scientific Chemicals

Catalog number: L20177.03
1 g, Each
Quantity
Catalog number: L20177.03
also known as L20177-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74181-34-3
IUPAC Name
2,2-dimethyl-1,3-dioxan-5-one
Molecular Formula
C6H10O3
InChI Key
ASFQDNDZFGFMMP-UHFFFAOYSA-N
SMILES
CC1(C)OCC(=O)CO1
Form
liquid
Refractive Index
1.4275 - 1.4355 @20?C
Appearance (Color)
clear colourless to pale yellow
Assay (GC)
> 88.0%

Description

2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethyl-1,3-dioxan-5-one is used in the preparation of 2,2-dimethyl-[1,3]dioxan-5-ol by reducing with lithium aluminum hydride. It is also used in the annulation of bete-(hetero)aryl-alfa-nitro-alfa,beta-enals.

Solubility
Miscible with water and most organic solvents.

Notes
Incompatible with oxidizing agents. Air, moisture and heat sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text