Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.
Solubility
Miscible with organic solvents, terahydro furan, dichloromethane and acetonitrile.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Chen, W.; Bay, W. P.; Wong, M. W.; Huang, D. Selenium Blue-alpha and -beta: turning on the fluorescence of a pyrenyl fluorophore via oxidative cleavage of the Se-C bond by reactive oxygen species. Tetrahedron Lett. 2012, 53 (30), 3843-3846.
Guillemin, J. C. Organic Selenocyanates: Synthesis, Characterization and Uses in Chemistry and Biology. Curr. Org. Chem. 2011, 15 (11), 1670-1687.