Phenyl selenocyanate, 98%, Thermo Scientific Chemicals
Phenyl selenocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenyl selenocyanate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L20194.06
also known as L20194-06
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
Add to cart
Phenyl selenocyanate, 98%, Thermo Scientific Chemicals
Catalog numberL20194.06
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2179-79-5
IUPAC Name(phenylselanyl)formonitrile
Molecular FormulaC7H5NSe
InChI KeyNODWRXQVQYOJGN-UHFFFAOYSA-N
SMILESN#C[Se]C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6010-1.6050 @ 20?C
Appearance (Color)Clear orange
Assay (GC)≥97.5%
FormLiquid
Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.

Solubility
Miscible with organic solvents, terahydro furan, dichloromethane and acetonitrile.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Chen, W.; Bay, W. P.; Wong, M. W.; Huang, D. Selenium Blue-alpha and -beta: turning on the fluorescence of a pyrenyl fluorophore via oxidative cleavage of the Se-C bond by reactive oxygen species. Tetrahedron Lett. 2012, 53 (30), 3843-3846.
  2. Guillemin, J. C. Organic Selenocyanates: Synthesis, Characterization and Uses in Chemistry and Biology. Curr. Org. Chem. 2011, 15 (11), 1670-1687.