Thermo Scientific Chemicals

Quinoline-4-carboxaldehyde, 97+%, Thermo Scientific Chemicals

Catalog number: L20203.14
25 g, Each
Thermo Scientific Chemicals

Quinoline-4-carboxaldehyde, 97+%, Thermo Scientific Chemicals

Catalog number: L20203.14
25 g, Each
Quantity
Catalog number: L20203.14
also known as L20203-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4363-93-3
IUPAC Name
quinoline-4-carbaldehyde
Molecular Formula
C10H7NO
InChI Key
MGCGJBXTNWUHQE-UHFFFAOYSA-N
SMILES
O=CC1=C2C=CC=CC2=NC=C1
Assay (GC)
≥97.0%
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Identification (FTIR)
Conforms
Appearance (Color)
White to cream or pale yellow
Free acid (titration)
≤1.5%

Description

4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Quinolinecarboxaldehyde(cinchoninaldehyde) was used in the synthesis of lepidylamines.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store under dry inert gas. Incompatible with oxidizing agents and air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text