Thermo Scientific Chemicals

Dysprosium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: L20252.14
25 g, Each
Thermo Scientific Chemicals

Dysprosium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: L20252.14
25 g, Each
Quantity
Catalog number: L20252.14
also known as L20252-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
139177-62-1
IUPAC Name
dysprosium(3+) tritrifluoromethanesulfonate
Molecular Formula
C3DyF9O9S3
InChI Key
XSVCYDUEICANRJ-UHFFFAOYSA-K
SMILES
[Dy+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Form
powder
Elemental Analysis
Fluorine F: 27.34 - 28.74% (Theory 28.04%)
Appearance (Color)
White
Assay (unspecified)
> 97.5% by Fluorine EA

Description

Dysprosium(III) trifluoromethanesulfonate is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes. This is an effective catalyst for electrophilic substitution reactions of indoles with imines. Also used as catalyst for Aza-Piancatelli rearrangement, Friedel-Crafts alkylation, ring-opening polymerization reactions, microwave-assisted Kabachnik-Fields condensation, cycloaddition reactions (Lewis-acid catalyst), fries rearrangement and enantioselective glyoxalate-ene reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dysprosium(III) trifluoromethanesulfonate is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes. This is an effective catalyst for electrophilic substitution reactions of indoles with imines. Also used as catalyst for Aza-Piancatelli rearrangement, Friedel-Crafts alkylation, ring-opening polymerization reactions, microwave-assisted Kabachnik-Fields condensation, cycloaddition reactions (Lewis-acid catalyst), fries rearrangement and enantioselective glyoxalate-ene reactions.

Solubility
Soluble in water.

Notes
Hygroscopic. store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text