Thermo Scientific Chemicals

Samarium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: L20253.06
5 g, Each
Thermo Scientific Chemicals

Samarium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: L20253.06
5 g, Each
Quantity
Catalog number: L20253.06
also known as L20253-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52093-28-4
IUPAC Name
samarium(3+) tritrifluoromethanesulfonate
Molecular Formula
C3F9O9S3Sm
InChI Key
DDCWGUIPLGMBPO-UHFFFAOYSA-K
SMILES
[Sm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Appearance (Color)
White
Form
Powder
Assay (unspecified)
≥97.5% (U.S. specification)
Assay from Suppliers CofA
≥97.5% based on elemental Fluorine (non-U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

Used as catalyst for Baylis-Hillman reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as catalyst for Baylis-Hillman reaction.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in a cool, dry place in a tightly closed containers. Stable under recommended storage conditions. Incompatible with oxidizing agents and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text