Thermo Scientific Chemicals

Cerium(IV) sulfate, anhydrous, 97%, Thermo Scientific Chemicals

Catalog number: L20254.14
25 g, Each
Thermo Scientific Chemicals

Cerium(IV) sulfate, anhydrous, 97%, Thermo Scientific Chemicals

Catalog number: L20254.14
25 g, Each
Quantity
Catalog number: L20254.14
also known as L20254-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13590-82-4
IUPAC Name
λ⁴-cerium(4+) disulfate
Molecular Formula
CeO8S2
InChI Key
VZDYWEUILIUIDF-UHFFFAOYSA-J
SMILES
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Assay from Suppliers CofA
≥96.0% (REO)
Form
Powder
Impurity content
Report Metals Analysis

Description

Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions' reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions′ reactions.

Solubility
Soluble in water and dilute sulfuric acid.

Notes
Store in cool place. Hygroscopic and moisture sensitive. Avoid moisture. Incompatible with strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text