Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate, 97%, ee 98+%, Thermo Scientific Chemicals
Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate, 97%, ee 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate, 97%, ee 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L20259.14
also known as L20259-14
Price (USD)/ Each
320.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
377.00 
Save 56.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
320.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
377.00 
Save 56.35 (15%)
Add to cart
Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate, 97%, ee 98+%, Thermo Scientific Chemicals
Catalog numberL20259.14
Price (USD)/ Each
320.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
377.00 
Save 56.35 (15%)
-
Add to cart
Chemical Identifiers
CAS312745-91-8
IUPAC Nameethyl 4-cyano-3-hydroxybutanoate
Molecular FormulaC7H11NO3
InChI KeyLOQFROBMBSKWQY-UHFFFAOYNA-N
SMILESCCOC(=O)CC(O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
Refractive Index1.4460-1.4510 @ 20?C
FormLiquid
Optical Rotation4.5? ± 0.5? (neat)
View more
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Zhong-Yu You; Zhi-Qiang Liu; Yu-Guo Zheng. Chemical and enzymatic approaches to the synthesis of optically pure ethyl (R)-4-cyano-3-hydroxybutanoate. Applied Microbiology and Biotechnology. 2014, 98 (1), 11-21.