6-Methoxy-2-naphthylglyoxal hydrate, 98%, dry wt. basis, Thermo Scientific Chemicals
6-Methoxy-2-naphthylglyoxal hydrate, 98%, dry wt. basis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methoxy-2-naphthylglyoxal hydrate, 98%, dry wt. basis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L20260.06
also known as L20260-06
Price (USD)/ Each
271.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
271.00
Add to cart
6-Methoxy-2-naphthylglyoxal hydrate, 98%, dry wt. basis, Thermo Scientific Chemicals
Catalog numberL20260.06
Price (USD)/ Each
271.00
-
Add to cart
Chemical Identifiers
CAS745783-88-4
IUPAC Name2-(6-methoxynaphthalen-2-yl)-2-oxoacetaldehyde
Molecular FormulaC13H10O3
InChI KeyBAVQWNGBJSPFBJ-UHFFFAOYSA-N
SMILESCOC1=CC2=CC=C(C=C2C=C1)C(=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Assay (GC)≥97.5%
Melting Point (clear melt)121.0-130.5?C
Water Content (Karl Fischer Titration)≤20.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only