Thermo Scientific Chemicals

3-Methoxyphenylglyoxal hydrate, 97%, dry wt. basis, Thermo Scientific Chemicals

Catalog number: L20263.03
1 g, Each
Thermo Scientific Chemicals

3-Methoxyphenylglyoxal hydrate, 97%, dry wt. basis, Thermo Scientific Chemicals

Catalog number: L20263.03
1 g, Each
Quantity
Catalog number: L20263.03
also known as L20263-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32025-65-3
IUPAC Name
2-(3-methoxyphenyl)-2-oxoacetaldehyde
Molecular Formula
C9H8O3
InChI Key
SIHYQEYAJMDKQH-UHFFFAOYSA-N
SMILES
COC1=CC=CC(=C1)C(=O)C=O
Assay (GC)
≥96.0% (dry wt. basis)
Appearance (Color)
Yellow to pale brown
Water Content (Karl Fischer Titration)
7-22% (ca. 1.5 waters)
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text