Thermo Scientific Chemicals

2,4-Difluorophenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals

Catalog number: L20266.06
5 g, Each
Thermo Scientific Chemicals

2,4-Difluorophenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals

Catalog number: L20266.06
5 g, Each
Quantity
Catalog number: L20266.06
also known as L20266-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79784-36-4
IUPAC Name
2-(2,4-difluorophenyl)-2-oxoacetaldehyde
Molecular Formula
C8H4F2O2
InChI Key
AZBQYELTMQNVST-UHFFFAOYSA-N
SMILES
FC1=CC(F)=C(C=C1)C(=O)C=O
Appearance (Color)
White to pale pink
Assay (GC)
≥94.0%
Melting Point (clear melt)
48.5-57.5?C
Form
Powder

Description

Used in the synthesis of biologically active 1,2,4-triazine derivatives. Also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of biologically active 1,2,4-triazine derivatives. Also used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text