Thermo Scientific Chemicals

3,4-Difluorophenylglyoxal hydrate, 98%, dry wt. basis, Thermo Scientific Chemicals

Catalog number: L20271.03
1 g, Each
Thermo Scientific Chemicals

3,4-Difluorophenylglyoxal hydrate, 98%, dry wt. basis, Thermo Scientific Chemicals

Catalog number: L20271.03
1 g, Each
Quantity
Catalog number: L20271.03
also known as L20271-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79784-34-2
IUPAC Name
2-(3,4-difluorophenyl)-2-oxoacetaldehyde
Molecular Formula
C8H4F2O2
InChI Key
VZRYIZGMQICGSF-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1F)C(=O)C=O
Appearance (Color)
Pale orange to pink
Assay (GC)
≥97.5%
Form
Powder
Water Content (Karl Fischer Titration)
≤17.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text