Thermo Scientific Chemicals

3,4-(Methylenedioxy)phenylglyoxal hydrate, 97%, dry wt basis, Thermo Scientific Chemicals

Catalog number: L20273.06
5 g, Each
Thermo Scientific Chemicals

3,4-(Methylenedioxy)phenylglyoxal hydrate, 97%, dry wt basis, Thermo Scientific Chemicals

Catalog number: L20273.06
5 g, Each
Quantity
Catalog number: L20273.06
also known as L20273-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65709-23-1
IUPAC Name
2-(2H-1,3-benzodioxol-5-yl)-2-oxoacetaldehyde
Molecular Formula
C9H6O4
InChI Key
OUCBKIWZWGSDJL-UHFFFAOYSA-N
SMILES
O=CC(=O)C1=CC2=C(OCO2)C=C1
Form
Powder or lumps
Melting Point (clear melt)
126.0-135.0?C
Appearance (Color)
Cream to pale brown or pale yellow
Assay (GC)
≥96.0% (dry wt. basis)
Water Content (Karl Fischer Titration)
5-15% (c. 0.5-1.5 waters)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text