Thermo Scientific Chemicals

Di-tert-butyl iminodicarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: L20325.14
25 g, Each
Thermo Scientific Chemicals

Di-tert-butyl iminodicarboxylate, 98%, Thermo Scientific Chemicals

Catalog number: L20325.14
25 g, Each
Quantity
Catalog number: L20325.14
also known as L20325-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51779-32-9
IUPAC Name
tert-butyl N-[(tert-butoxy)carbonyl]carbamate
Molecular Formula
C10H19NO4
InChI Key
XCAQIUOFDMREBA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NC(=O)OC(C)(C)C
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%

Description

Di-tert-butyl-iminodicarboxylate may be used in the preparation of trans-diamino-2-butene and cis-1,4-diamino-2-butene; di-tert-butyl N-3-butenyliminodicarboxylate; N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate & C1-C20 and C21-C40 fragments of tetrafibricin. Modified Gabriel Reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butyl-iminodicarboxylate may be used in the preparation of trans-diamino-2-butene and cis-1,4-diamino-2-butene; di-tert-butyl N-3-butenyliminodicarboxylate; N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate & C1-C20 and C21-C40 fragments of tetrafibricin. Modified Gabriel Reagent.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text