Thermo Scientific Chemicals

(S)-(-)-3-Butyn-2-ol, 99%, Thermo Scientific Chemicals

Catalog number: L20345.06
5 g, Each
Thermo Scientific Chemicals

(S)-(-)-3-Butyn-2-ol, 99%, Thermo Scientific Chemicals

Catalog number: L20345.06
5 g, Each
Quantity
Catalog number: L20345.06
also known as L20345-06
Price (USD)
Quantity
-
Have Questions?

Description

(S)-(-)-3-Butyn-2-ol is used to prepare (S)-(-)-2-tert-butyldimethylsiloxybut-3-yne. It finds application in ruthenium-catalyzed polymerization to get helical polyacetylenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Butyn-2-ol is used to prepare (S)-(-)-2-tert-butyldimethylsiloxybut-3-yne. It finds application in ruthenium-catalyzed polymerization to get helical polyacetylenes.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text