1-Methoxy-2-methyl-2-propanol, 98+%, Thermo Scientific Chemicals
1-Methoxy-2-methyl-2-propanol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methoxy-2-methyl-2-propanol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L20351.14
also known as L20351-14
Price (USD)/ Each
251.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
251.00
Add to cart
1-Methoxy-2-methyl-2-propanol, 98+%, Thermo Scientific Chemicals
Catalog numberL20351.14
Price (USD)/ Each
251.00
-
Add to cart
Chemical Identifiers
CAS3587-64-2
IUPAC Name1-methoxy-2-methylpropan-2-ol
Molecular FormulaC5H12O2
InChI KeyMXUXZWFVAPTPAG-UHFFFAOYSA-N
SMILESCOCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
Refractive Index1.4030-1.4070 @ 20?C
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only