Thermo Scientific Chemicals

Methyl potassium malonate, 98%, Thermo Scientific Chemicals

Catalog number: L20429.14
25 g, Each
Thermo Scientific Chemicals

Methyl potassium malonate, 98%, Thermo Scientific Chemicals

Catalog number: L20429.14
25 g, Each
Quantity
Catalog number: L20429.14
also known as L20429-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38330-80-2
IUPAC Name
potassium 3-methoxy-3-oxopropanoate
Molecular Formula
C4H5KO4
InChI Key
WWTULTKUWBKVGV-UHFFFAOYSA-M
SMILES
[K+].COC(=O)CC([O-])=O
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.5%

Description

Methyl potassium malonate is a protein advanced glycosylation inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl potassium malonate is a protein advanced glycosylation inhibitor.

Solubility
Soluble in methanol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text