1,3-Bis(4-piperidinyl)propane, 97+%, Thermo Scientific Chemicals
1,3-Bis(4-piperidinyl)propane, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Bis(4-piperidinyl)propane, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L20442.14
also known as L20442-14
Price (USD)/ Each
37.40
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
37.40
Add to cart
1,3-Bis(4-piperidinyl)propane, 97+%, Thermo Scientific Chemicals
Catalog numberL20442.14
Price (USD)/ Each
37.40
-
Add to cart
Chemical Identifiers
CAS16898-52-5
IUPAC Name4-[3-(piperidin-4-yl)propyl]piperidine
Molecular FormulaC13H26N2
InChI KeyOXEZLYIDQPBCBB-UHFFFAOYSA-N
SMILESC(CC1CCNCC1)CC1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.0%
Appearance (Color)White to cream or pale yellow
Formcrystalline powder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only