Invitrogen™

Lipofectamine™ 3000 Transfection Reagent

Catalog number: L3000150
Have Questions?
Invitrogen™

Lipofectamine™ 3000 Transfection Reagent

Catalog number: L3000150
Have Questions?
Catalog Number
L3000150
Unit Size
15 ml
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
0.1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
0.75 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1.5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 1.5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
L300015015 ml
-
L30000010.1 mL
-
L30000080.75 mL
-
L30000151.5 mL
-
L30000755 x 1.5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Lipofectamine™ 3000 Transfection Reagent leverages our most advanced lipid nanoparticle technology to provide superior transfection performance with improved application outcomes and reproducible results. This reagent delivers superior transfection efficiency and improved cell viability for the widest range of hard-to-transfect and common cells (e.g., HEK293, HeLa).

Successfully transfect the widest variety of biologically relevant cell types with a reagent that offers:

• Superior performance—our highest efficiency reagent for difficult-to-transfect cells
• Improved cell viability—gentle on your cells, with low toxicity
• Versatile—one reagent for DNA, RNA, and co-transfection

Efficiently Transfect Difficult-to-Transfect Cells
Lipofectamine™ 3000 reagent is designed to efficiently transfect difficult-to-transfect cells, yielding superior transfection performance across the broadest array of cell types. Successfully transfected cell types include:

Fibroblast (3T3, COS-7)
Myoblast (C2C12, L6 CRL-1458)
Hepatocyte (HepG2, HuH7)
Erythroleukemic cell (K562)
Breast carcinoma (MCF7, Hs578T)
Prostate carcinoma (LNCap)
Lung carcinoma (A549, NCI-H460)
Osteosarcoma (U-2 OS, Saos-2)
Colon carcinoma (Caco2, SW480)
Pancreatic carcinoma (PANC-1)
Skin melanoma (SK-MEL-28)

View performance data for 60+ cell lines plus comparison to Lipofectamine 2000 >

Transfect Common Cells Efficiently and Cost-Effectively
For common cell types, Lipofectamine™ 3000 reagent is more cost-effective than other reagents due to its high concentration and low usage amount. The 1.5 mL size provides up to 1500 transfection reactions (in 24-well plates).

Achieve Superior Transfection Efficiency While Maintaining Low Toxicity
Lipofectamine™ 3000 reagent maintains a high transfection efficiency within a robust dynamic range of lipid doses for quick and easy optimization. A low toxicity lipid dose (0.75 μL for 24-well plates) is suggested for applications requiring minimal disruption of the cells.

Enable High Transfection Efficiency in Novel Genome Editing Applications
Lipofectamine™ 3000 reagent is suitable for novel genome editing applications. It increases the likelihood of successful cleavage and recombination with TALENs or CRISPRs through highly efficient transfection, ultimately maximizing the efficiency of genetic modifications and simplifying the downstream processes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Classification
Animal Origin-Free
Cell Type
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type
Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Transfection Technique
Lipid-based Transfection
For Use With (Application)
Transfection
Green Features
Sustainable packaging
Product Line
Lipofectamine™
Product Type
Transfection Reagent
Quantity
15 mL
Serum Compatible
Yes

Contents & Storage

Contains one vial of Lipofectamine™ 3000 Reagent and one vial of P3000TM Enhancer Reagent. Store at 2–8°C. Do not freeze.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text