1x1 image pixel for data collection
Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), fluorescein (FITC)
Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), fluorescein (FITC)
Invitrogen™

Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), fluorescein (FITC)

Lycopersicon esculentum (Tomato) Lectin (LEL, TL), fluorescein (FITC), is a bright-green LEL-fluorophore conjugate. The tomato lectin LEL is widely usedRead more
Promo ImagePromotion
Promo code:RIRP25
Save on Imaging Essentials to Create Your Next Masterpiece
14% off your purchase of $499 or more on Invitrogen cell stains and mounting products Learn More
Have Questions?
Catalog NumberQuantity
L32478Promo Image1 mg
Catalog number L32478
Price (USD)
214.65
Online Exclusive
222.00
Save 7.35 (3%)
Each
Add to cart
Quantity:
1 mg
Price (USD)
214.65
Online Exclusive
222.00
Save 7.35 (3%)
Each
Add to cart
Lycopersicon esculentum (Tomato) Lectin (LEL, TL), fluorescein (FITC), is a bright-green LEL-fluorophore conjugate. The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research. LEL-FITC can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.

View complete list of fluorescent dye-conjugated lectins ›

• Very bright FITC conjugate detected with traditional FITC filter set, Ex/Em: 495/515 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
• Inhibiting/eluting sugar: chitin hydrolysate
• Blood group specificity: non-specific
• Unconjugated lectin molecular weight: 71 KDa.
• Immunofluorescence working concentration: 5–20 μg/mL

LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeClassic Dyes
Protein SubtypeLectins
Quantity1 mg
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
SourceTomato
ConjugateFluorescein
FormSolution
Unit SizeEach
Contents & Storage
When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept.

Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link