Invitrogen™

LIVE/DEAD™ Sperm Viability Kit

Catalog number: L7011
Have Questions?
Invitrogen™

LIVE/DEAD™ Sperm Viability Kit

Catalog number: L7011
Have Questions?
Catalog Number
L7011
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
L70111 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The LIVE/DEAD Sperm Viability Kit provides a novel fluorescence-based assay for analyzing the viability and fertilizing potential of sperm by fluorescence microscopy or flow cytometry. Membrane-permeant SYBR® 14 nucleic acid stain labels live sperm with green fluorescence, and membrane-impermeant propidium iodide labels the nucleic acids of membrane-compromised sperm with red fluorescence.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Eukaryotic Cells, Sperm Cells
Color
Green, Red
Description
LIVE/DEAD™ Sperm Viability Kit
Detection Method
Fluorescence
For Use With (Application)
Viability Assay
For Use With (Equipment)
Fluorescence Microscope, Flow Cytometer
Product Type
Sperm Viability Kit
Dye Type
Other Label(s) or Dye(s)
Emission
516, 617 nm
Excitation Wavelength Range
Visible light
Format
Tube(s), Slide(s)
Product Line
LIVE/DEAD™
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer at -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text