Invitrogen™

LDS 751

Catalog number: L7595
Have Questions?
Invitrogen™

LDS 751

Catalog number: L7595
Have Questions?
Catalog Number
L7595
Unit Size
10 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
L759510 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
LDS 751 is a cell-permeant nucleic acid stain that has a peak excitation at ∼543 nm on dsDNA and is particular useful in multicolor analyses due to its long wavelength emission maximum (∼712 nm). Binding of LDS 751 to dsDNA results in a ∼20-fold fluorescence enhancement.

View a selection guide for all Cell Viaibility Assays for Flow Cytometry.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Excitation Wavelength Range
543⁄712
Dye Type
LDS 751
For Use With (Equipment)
Flow Cytometer
Quantity
10 mg
Detection Method
Fluorescence
Form
Solid
Shipping Condition
Room Temperature
Product Type
Nucleic Acid Stain
SubCellular Localization
Nucleic Acids, Nucleus
Unit Size
10 mg

Contents & Storage

Contains 1 vial of LDS 751 (10 mg). Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text